Skip to Content
Merck
All Photos(2)

Key Documents

8.22254

Sigma-Aldrich

Formic acid

for synthesis

Synonym(s):

Methanoic acid, Formylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€50.90
2.5 L
€97.70
25 L
€314.00

€50.90


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€50.90
2.5 L
€97.70
25 L
€314.00

About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-579-1
NACRES:
NA.22

€50.90


Estimated to ship on05 May 2025Details


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

Assay

≥99% (acidimetric)

form

liquid

autoignition temp.

1004 °F

expl. lim.

12-38 % (v/v)
57 %

reaction suitability

reagent type: reductant

technique(s)

electrophoresis: suitable

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 10 g/L in H2O)

bp

100-101 °C (lit.)
101 °C/1013 hPa

mp

8.2-8.4 °C (lit.)

transition temp

flash point 49.5 °C

density

1.22 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Atmospheric Implications: Provides insights into the large and ubiquitous source of atmospheric formic acid, affecting precipitation chemistry and acidity (Millet et al., 2015).
  • Sustainable Production: Covers the sustainability aspects of formic acid production and its role in bridging power and chemical industries (Bulushev & Ross, 2018).
  • Hydrogen Energy Carrier: Explores formic acid as a potential hydrogen storage medium, focusing on its applications in low-carbon energy systems (Singh et al., 2016).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Density (d 20 °C/ 4 °C): 1.218 - 1.221
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

121.1 °F - closed cup

Flash Point(C)

49.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service