Skip to Content
Merck
All Photos(1)

Key Documents

8.22188

Sigma-Aldrich

TWEEN® 85

for synthesis

Synonym(s):

Polyoxyethylene sorbitan trioleate

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€66.20

€66.20


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 ML
€66.20

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.22

€66.20


Available to ship on31 March 2025Details


Request a Bulk Order

vapor pressure

1.4 hPa ( 20 °C)

Quality Level

form

liquid

potency

>36000 mg/kg LD50, oral (Rat)

transition temp

flash point >149 °C (DIN 51758)

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Density (d 20 °C/ 4 °C): 1.028 - 1.033
Hydroxyl value: 39 - 52
Saponification value: 80 - 95
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

300.2 °F - DIN 51758

Flash Point(C)

> 149 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service