Skip to Content
Merck
All Photos(1)

Key Documents

8.22140

Sigma-Aldrich

1,4-Dihydro-4-pyridinone

for synthesis

Synonym(s):

1,4-Dihydro-4-pyridinone, 4(1H)-Pyridone

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€252.00

€252.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
€252.00

About This Item

Empirical Formula (Hill Notation):
C5H5NO
CAS Number:
Molecular Weight:
95.10
MDL number:
UNSPSC Code:
12352115
EC Index Number:
203-633-2
NACRES:
NA.22

€252.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Quality Level

form

solid

autoignition temp.

515 °C

potency

4600 mg/kg LD50, oral (Rat)

pH

7.0 (20 °C, 50 g/L in H2O)

bp

170-172 °C/0.03 hPa

mp

140-147 °C

transition temp

flash point 212 °C

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1cc[c](cc1)=O

InChI

1S/C5H5NO/c7-5-1-3-6-4-2-5/h1-4H,(H,6,7)

InChI key

GCNTZFIIOFTKIY-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 95.0 %
Sulfated ash: ≤ 5.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

413.6 °F

Flash Point(C)

212 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service