Skip to Content
Merck
All Photos(1)

Key Documents

8.22028

Sigma-Aldrich

1,8-Dihydroxyanthraquinone

for synthesis

Synonym(s):

1,8-Dihydroxyanthraquinone, Chrysazin, Dantron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-173-5
form:
powder
Assay:
≥97.0% (HPLC)

Quality Level

Assay

≥97.0% (HPLC)

form

powder

mp

190-193 °C

transition temp

flash point >200 °C

bulk density

600 kg/m3

storage temp.

2-30°C

InChI

1S/C14H8O4/c15-9-5-1-3-7-11(9)14(18)12-8(13(7)17)4-2-6-10(12)16/h1-6,15-16H

InChI key

QBPFLULOKWLNNW-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 190 °C
Melting range (upper value): ≤ 194 °C
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service