Skip to Content
Merck
All Photos(1)

Key Documents

8.21762

Sigma-Aldrich

2,5-Di-tert-butylhydroquinone

for synthesis

Synonym(s):

2,5-Di-tert-butylhydroquinone

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€45.10
1 KG
€158.00
25 KG
€1,760.00

€45.10


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€45.10
1 KG
€158.00
25 KG
€1,760.00

About This Item

Linear Formula:
2,5-[CH3)3C]2C6H2-1,4-(OH)2
CAS Number:
Molecular Weight:
222.32
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-841-8
NACRES:
NA.22

€45.10


Estimated to ship on30 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

autoignition temp.

420 °C

mp

212-218 °C

storage temp.

2-30°C

SMILES string

Oc1c(cc(c(c1)C(C)(C)C)O)C(C)(C)C

InChI

1S/C14H22O2/c1-13(2,3)9-7-12(16)10(8-11(9)15)14(4,5)6/h7-8,15-16H,1-6H3

InChI key

JZODKRWQWUWGCD-UHFFFAOYSA-N

Application

  • Food Contact Material Study: Investigated the occurrence and application areas of 2,5-di-tert-butylhydroquinone in food contact materials, highlighting its use and safety implications (Pedersen & Cederberg, 2014).
  • Antibacterial and Anti-Biofilm Properties: Explored the potential use of DBHQ as an antibacterial and anti-biofilm agent against Staphylococcus aureus, suggesting new applications in medical and healthcare fields (Kim et al., 2022).
  • Cognitive Performance and Metabolomics: Investigated serum and CSF metabolites, including DBHQ, in stroke-free patients, offering insights into vascular risk factors and cognitive performance, relevant for neurological drug development (Peng et al., 2020).
  • Cellular Mechanism Study: Assessed the role of DBHQ in Ca2+ handling in cellular models, which could be critical for understanding cellular mechanisms in drug discovery and development (Liu et al., 2015).
  • Review on Food Additives: A narrative review highlighting the excessive use of DBHQ in food products and its potential health impacts, emphasizing the need for accurate monitoring and regulatory review (Khezerlou et al., 2022).

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 212 °C
Melting range (upper value): ≤ 218 °C
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service