Skip to Content
Merck
All Photos(1)

Key Documents

8.21177

Sigma-Aldrich

Trimethylamine

(40% solution in water) for synthesis

Synonym(s):

N,N,N-Trimethylamine, N,N-Dimethylmethanamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€37.70
1 L
€43.10
2.5 L
€107.00
21 KG
€628.00

€37.70


Estimated to ship on12 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€37.70
1 L
€43.10
2.5 L
€107.00
21 KG
€628.00

About This Item

UNSPSC Code:
12352116
NACRES:
NA.22

€37.70


Estimated to ship on12 May 2025Details


Request a Bulk Order

vapor pressure

600 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

250 °C

potency

500 mg/kg LD50, oral (Rat)

expl. lim.

2-11.6 % (v/v) anhydrous substance)

pH

13 (20 °C in H2O)

bp

30.8 °C

mp

-2 °C

transition temp

flash point -45 °C

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

Application


  • Blood trimethylamine-N-oxide origins: Explores the microbiota-mediated breakdown of phosphatidylcholine and absorption from the small intestine, significant for drug discovery in cardiovascular diseases (Stremmel et al., 2017).

  • Diabetes and trimethylamine N-oxide levels: Discusses the strong associations between systemic trimethylamine N-oxide levels and diabetes, offering insights into potential therapeutic targets for drug discovery (Dambrova et al., 2016).

  • Detection via lanthanide metal-organic framework: Trimethylamine-N-oxide detection using a lanthanide MOF, critical for advancing sensor technology in material science (Min et al., 2022).

  • Trimethylamine N-Oxide research: Overview of trimethylamine N-oxide′s roles, focusing on dietary sources and health implications, valuable for academia and nutritional studies (Velasquez et al., 2016).

  • Efficient gas sensors: Application of MoS2/SnO2 heterojunctions in creating effective gas sensors for trimethylamine, useful in material science for developing new sensor technologies (Qiao et al., 2018).


Analysis Note

Assay (acidimetric): 38.0 - 41.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-49.0 °F

Flash Point(C)

-45 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service