Skip to Content
Merck
All Photos(1)

Key Documents

8.20959

Sigma-Aldrich

Tetraethylene glycol dimethyl ether

for synthesis

Synonym(s):

Tetraethylene glycol dimethyl ether, Dimethyl tetraglycol, Bis[2-(2-methoxyethoxy)ethyl]ether

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€51.80
1 L
€149.00

€51.80


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
€51.80
1 L
€149.00

About This Item

Linear Formula:
CH3O(CH2CH2O)4CH3
CAS Number:
Molecular Weight:
222.28
MDL number:
UNSPSC Code:
12352112
EC Index Number:
205-594-7
NACRES:
NA.22

€51.80


Estimated to ship on22 April 2025Details


Request a Bulk Order

vapor pressure

0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

195 °C (DIN 51794)

potency

3850 mg/kg LD50, oral (Rat)

expl. lim.

0.8 % (v/v)

kinematic viscosity

4.1 cSt(20 °C)

bp

272-277 °C/1013 hPa

mp

-30 °C

transition temp

flash point 141 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOCCOC)CCOCCOC

InChI

1S/C10H22O5/c1-11-3-5-13-7-9-15-10-8-14-6-4-12-2/h3-10H2,1-2H3

InChI key

ZUHZGEOKBKGPSW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.010 - 1.012
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service