Skip to Content
Merck
All Photos(1)

Key Documents

8.20809

Sigma-Aldrich

1-Methylnaphthalene

for synthesis

Synonym(s):

1-Methylnaphthalene

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€25.70
250 ML
€57.70
1 L
€183.00

€25.70


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
5 ML
€25.70
250 ML
€57.70
1 L
€183.00

About This Item

Empirical Formula (Hill Notation):
C11H10
CAS Number:
Molecular Weight:
142.20
MDL number:
UNSPSC Code:
12352005
EC Index Number:
201-966-8
NACRES:
NA.22

€25.70


Estimated to ship on31 March 2025Details


Request a Bulk Order

vapor pressure

2 hPa ( 25 °C)

Quality Level

Assay

≥94.0% (GC)

form

liquid

autoignition temp.

529 °C

potency

1840 mg/kg LD50, oral (Rat)

expl. lim.

0.7-6.5 % (v/v)

bp

244-245 °C/1013 mbar

mp

<-30 °C

transition temp

flash point 82 °C

solubility

0.026 g/L

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Cc1c2c(ccc1)cccc2

InChI

1S/C11H10/c1-9-5-4-7-10-6-2-3-8-11(9)10/h2-8H,1H3

InChI key

QPUYECUOLPXSFR-UHFFFAOYSA-N

Application

  • Solvent-assisted microextraction: 1-Methylnaphthalene was used in the development of sponge-like porous manganese(II, III) oxide coatings for solvent-assisted solid-phase microextraction, demonstrating its utility in environmental monitoring and analysis of polycyclic aromatic hydrocarbons (Saraji and Nobakht, 2022).
  • Catalytic applications: As a tar-type compound, 1-Methylnaphthalene was investigated for its degradation under mild conditions using TiO2-supported Pd catalysts, emphasizing its importance in industrial applications for cleaner production technologies (Raad et al., 2021).
  • Advanced oxidation processes: Research involving nano-MoO2 and peroxymonosulfate demonstrated the effective degradation of PAH derivatives, including 1-Methylnaphthalene, underscoring its relevance in water treatment technologies and pollution control (Chen et al., 2021).

Analysis Note

Assay (GC, area%): ≥ 94.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.015 - 1.025
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service