Skip to Content
Merck
All Photos(1)

Key Documents

8.20760

Sigma-Aldrich

di-Manganese decacarbonyl

for synthesis

Synonym(s):

di-Manganese decacarbonyl, Decacarbonyldimanganese, Manganese carbonyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€110.00

€110.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
€110.00

About This Item

Linear Formula:
Mn2(CO)10
CAS Number:
Molecular Weight:
389.98
MDL number:
UNSPSC Code:
12161603
EC Index Number:
233-445-6
NACRES:
NA.22

€110.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Quality Level

form

crystals

mp

150 °C

storage temp.

2-8°C

SMILES string

[Mn].[Mn].O=[C].O=[C].O=[C].O=[C].O=[C].O=[C].O=[C].O=[C].O=[C].O=[C]

InChI

1S/10CO.2Mn/c10*1-2;;

InChI key

QFEOTYVTTQCYAZ-UHFFFAOYSA-N

Analysis Note

Assay (out of Mn): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service