Skip to Content
Merck
All Photos(1)

Key Documents

8.20709

Sigma-Aldrich

Isatin

for synthesis

Synonym(s):

Isatin

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€37.40
250 G
€133.00

€37.40


Estimated to ship on26 March 2025Details



Select a Size

Change View
5 G
€37.40
250 G
€133.00

About This Item

Empirical Formula (Hill Notation):
C8H5NO2
CAS Number:
Molecular Weight:
147.13
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-077-8
NACRES:
NA.22

€37.40


Estimated to ship on26 March 2025Details


Quality Level

Assay

≥98.0% (GC)

autoignition temp.

220 °C

pH

7 (20 °C, 1.9 g/L in H2O)

mp

199-204 °C

transition temp

flash point 200 °C

solubility

2 g/L

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

N1c2c(cccc2)C(=O)C1=O

InChI

1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)9-8(7)11/h1-4H,(H,9,10,11)

InChI key

JXDYKVIHCLTXOP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 199 °C
Melting range (upper value): ≤ 204 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service