Skip to Content
Merck
All Photos(2)

Key Documents

8.20692

Sigma-Aldrich

4′-Hydroxypropiophenone

for synthesis

Synonym(s):

4′-Hydroxypropiophenone

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€64.00

€64.00


Estimated to ship on28 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€64.00

About This Item

Linear Formula:
4-(OH)C6H4COC2H5
CAS Number:
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-743-2
NACRES:
NA.22

€64.00


Estimated to ship on28 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

potency

11800 mg/kg LD50, oral (Rat)

mp

148-151 °C

transition temp

flash point 112 °C

solubility

0.345 g/L

density

1.09 g/cm3 at 20 °C

bulk density

300‑400 kg/m3

storage temp.

2-30°C

SMILES string

Oc1ccc(cc1)C(=O)CC

InChI

1S/C9H10O2/c1-2-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

RARSHUDCJQSEFJ-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 148 °C
Melting range (upper value): ≤ 151 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service