Skip to Content
Merck
All Photos(1)

Key Documents

8.20678

Sigma-Aldrich

5-Hydroxymethyl-2-furancarbaldehyde

for synthesis

Synonym(s):

5-Hydroxymethyl-2-furancarbaldehyde, 5-(Hydroxymethyl)furfurol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€36.90
1 G
€65.00
5 G
€168.00

€36.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
€36.90
1 G
€65.00
5 G
€168.00

About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352114
EC Index Number:
200-654-9
NACRES:
NA.22

€36.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

solid

potency

2500 mg/kg LD50, oral (Rat)

bp

154-155 °C/16 hPa

mp

27-30 °C

transition temp

flash point 79 °C

storage temp.

2-8°C

SMILES string

[o]1c(ccc1C=O)CO

InChI

1S/C6H6O3/c7-3-5-1-2-6(4-8)9-5/h1-3,8H,4H2

InChI key

NOEGNKMFWQHSLB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 28 °C
Melting range (upper value): ≤ 33 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service