Skip to Content
Merck
All Photos(1)

Key Documents

8.20610

Sigma-Aldrich

Glyoxal

(40% solution in water) for synthesis

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€31.90
1 L
€60.00
2.5 L
€120.00
25 L
€614.00

€31.90


Estimated to ship on27 March 2025Details



Select a Size

Change View
100 ML
€31.90
1 L
€60.00
2.5 L
€120.00
25 L
€614.00

About This Item

UNSPSC Code:
12352114
NACRES:
NA.22

€31.90


Estimated to ship on27 March 2025Details


vapor pressure

20.2 hPa ( 20 °C)

Quality Level

autoignition temp.

285 °C

potency

2000-5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

2-3 (20 °C in H2O)

bp

104 °C/1013 hPa

mp

-14 °C

density

1.27 g/cm3 at 20 °C

storage temp.

2-30°C

Application


  • HFIP-Mediated Multicomponent Reactions: Research demonstrates the use of HFIP (Hexafluoroisopropanol) in synthesizing pyrazole-linked thiazole derivatives, highlighting a new application in organic synthesis. Glyoxal is instrumental in the development of these multifunctional structures, contributing to advanced material design (Banerjee et al., 2024).

  • Peptide Stapling in Protein Engineering: A study describes peptide stapling techniques using alpha-ketoaldehydes, which crosslink amines through glyoxal-modified linkers. This approach is pivotal in creating stable peptide structures for therapeutic applications (Guo et al., 2024).

Analysis Note

Assay (acidimetric): 39.0 - 41.0 %
Density (d 20 °C/ 4 °C): 1.266 - 1.271

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Muta. 2 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service