Skip to Content
Merck
All Photos(1)

Key Documents

8.20551

Sigma-Aldrich

1,2-Epoxybutane

for synthesis

Synonym(s):

1,2-Epoxybutane, Ethyloxirane, 1,2-Butylene oxide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€33.10
1 L
€61.00
2.5 L
€137.00

€33.10


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 ML
€33.10
1 L
€61.00
2.5 L
€137.00

About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-438-2
NACRES:
NA.22

€33.10


In StockDetails


Request a Bulk Order

vapor pressure

1005 hPa ( 63 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

370 °C

potency

500 mg/kg LD50, oral (Rat)
1743 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-19 % (v/v)

pH

7 (20 °C, 50 g/L in H2O)

bp

63 °C/1013 hPa

mp

-130 °C

transition temp

flash point -26 °C

solubility

86.8 g/L

density

0.83 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(C1)CC

InChI

1S/C4H8O/c1-2-4-3-5-4/h4H,2-3H2,1H3

InChI key

RBACIKXCRWGCBB-UHFFFAOYSA-N

Application

  • Efficient catalysis of CO2 cycloaddition: 1,2-Epoxybutane is utilized in the synthesis of hydroxy-rich covalent organic frameworks, proving effective in catalyzing CO2 cycloaddition. This application supports environmental sustainability efforts through the conversion of carbon dioxide into useful compounds (Pang et al., 2023).
  • Strain-release reagent synthesis: Highlighting its versatility as a chemical intermediate, 1,2-Epoxybutane is used in one-pot syntheses of strain-release reagents from methyl sulfones. This research opens new avenues in synthetic chemistry for creating novel organic compounds efficiently (Jung and Lindsay, 2022).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.830 - 0.831
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service