Skip to Content
Merck
All Photos(1)

Key Documents

8.20392

Sigma-Aldrich

1,3-Diethoxy-2-propanol

for synthesis

Synonym(s):

1,3-Diethoxy-2-propanol, Glycerol α,γ-diethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€72.20
250 ML
€269.00

€72.20


In StockDetails


Request a Bulk Order

Select a Size

Change View
50 ML
€72.20
250 ML
€269.00

About This Item

Linear Formula:
C2H5OCH2CH(OH)CH2OC2H5
CAS Number:
Molecular Weight:
148.20
MDL number:
UNSPSC Code:
12352104
EC Index Number:
223-736-6
NACRES:
NA.22

€72.20


In StockDetails


Request a Bulk Order

Quality Level

Assay

≥96.0% (GC)

form

liquid

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC(O)COCC)CC

InChI

1S/C7H16O3/c1-3-9-5-7(8)6-10-4-2/h7-8H,3-6H2,1-2H3

InChI key

WIHIUTUAHOZVLE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.950 - 0.960
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service