Skip to Content
Merck
All Photos(1)

Key Documents

8.20308

Sigma-Aldrich

4-(Hydroxymercuri)benzoic acid sodium salt

≥97%

Synonym(s):

4-(Hydroxymercuri)benzoic acid sodium salt, PCMB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HOHg)C6H4COONa
CAS Number:
Molecular Weight:
360.69
MDL number:
UNSPSC Code:
12352103
EC Index Number:
205-340-5
Pricing and availability is not currently available.

Assay

≥97%

form

powder

storage temp.

2-30°C

InChI

1S/C7H5O2.Hg.Na.H2O/c8-7(9)6-4-2-1-3-5-6;;;/h2-5H,(H,8,9);;;1H2/q;;+1;/p-1

InChI key

BERBQUNVJGVZCX-UHFFFAOYSA-M

General description

for synthesis

Analysis Note

Assay (ex Hg): ≥ 97.0 %
Identity (IR): passes test

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service