Skip to Content
Merck
All Photos(2)

Key Documents

8.20254

Sigma-Aldrich

(+)-Camphene

for synthesis

Synonym(s):

(+)-Camphene, 2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane, 3,3-Dimethyl-2-methylenenorcamphane

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€40.70
1 KG
€137.00

€40.70


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€40.70
1 KG
€137.00

About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
MDL number:
UNSPSC Code:
12352002
EC Index Number:
201-234-8
NACRES:
NA.22

€40.70


Estimated to ship on31 March 2025Details


Request a Bulk Order

vapor pressure

3.99 hPa ( 20 °C)

Quality Level

Assay

≥90.0% (GC)

form

crystals

autoignition temp.

265 °C

potency

>2500 mg/kg LD50, skin (Rabbit)

pH

5.5 (22 °C in H2O, saturated aqueous solution)

bp

158-159 °C/1013 hPa

mp

42-45 °C

transition temp

flash point 26 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C21CC(CC2)C(=C)C1(C)C

InChI

1S/C10H16/c1-7-8-4-5-9(6-8)10(7,2)3/h8-9H,1,4-6H2,2-3H3

InChI key

CRPUJAZIXJMDBK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 90.0 % (a/a)
Assay (GC, Tricyclene): ≤ 10.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

78.8 °F - DIN 51755 Part 1

Flash Point(C)

26 °C - DIN 51755 Part 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service