Skip to Content
Merck
All Photos(1)

Key Documents

8.20063

Sigma-Aldrich

2-Hydroxyethylammonium chloride

for synthesis

Synonym(s):

2-Hydroxyethylammonium chloride, Ethanolamine hydrochloride, 2-Aminoethanol hydrochloride, Ethanolammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€36.80
500 G
€64.70

€36.80


In StockDetails



Select a Size

Change View
100 G
€36.80
500 G
€64.70

About This Item

Empirical Formula (Hill Notation):
C2H7NO · xHCl
CAS Number:
Molecular Weight:
61.08 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
217-900-6
NACRES:
NA.22

€36.80


In StockDetails


Quality Level

mp

80-84 °C

storage temp.

2-30°C

SMILES string

Cl.NCCO

InChI

1S/C2H7NO.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

PMUNIMVZCACZBB-UHFFFAOYSA-N

General description

2-Hydroxyethylammonium chloride also known as ethanolammonium chloride used in the synthesis of esters and ethers via nucleophilic substitution reactions. It can be used to functionalization of various polymers.

Analysis Note

Assay (argentometric): ≥ 99.0 %
Melting range (lower value): ≥ 80 °C
Melting range (upper value): ≤ 84 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service