Skip to Content
Merck
All Photos(1)

Key Documents

8.20030

Sigma-Aldrich

Ethylenediamine monohydrate

for synthesis

Synonym(s):

Ethylenediamine monohydrate, 1,2-Diaminoethane, 1,2-Ethanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H8N2 · xH2O
CAS Number:
Molecular Weight:
60.10 (anhydrous basis)
UNSPSC Code:
12352116
EC Index Number:
203-468-6

vapor pressure

12 hPa ( 20 °C)

Assay

≥99% (acidimetric)

form

liquid

autoignition temp.

385 °C

potency

866 mg/kg LD50, oral (Rat)
560 mg/kg LD50, skin (Rabbit)

expl. lim.

2.7-16.6 % (v/v)

pH

12 (20 °C, 100 g/L in H2O)

bp

118 °C/1013 hPa

mp

8-10 °C

transition temp

flash point 42 °C

density

0.963 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C2H8N2.H2O/c3-1-2-4;/h1-4H2;1H2

InChI key

XZUAPPXGIFNDRA-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99
Identity (IR-spectrum): passes test
Density (d 20 °C/ 4 °C): 0.961 - 0.964

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(C)

42 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service