Skip to Content
Merck
All Photos(1)

Key Documents

8.18847

Sigma-Aldrich

Ethylene glycol dimethacrylate

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

Ethylene glycol dimethacrylate, Ethylene dimethacrylate, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€38.60
1 L
€150.00

€38.60


Estimated to ship on17 February 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
€38.60
1 L
€150.00

About This Item

Empirical Formula (Hill Notation):
C10H14O4
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-617-2
NACRES:
NA.22

€38.60


Estimated to ship on17 February 2025Details


Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥97.5% (GC)

form

liquid

potency

3300 mg/kg LD50, oral (Rat)

bp

240 °C/1013 hPa

mp

-20 °C

transition temp

flash point 101.5 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOC(=O)C(=C)C)C(=O)C(=C)C

InChI

1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3

InChI key

STVZJERGLQHEKB-UHFFFAOYSA-N

Application

  • Development of antioxidant and antimicrobial membranes: A study demonstrated the use of ethylene glycol dimethacrylate in the crosslinking of chitosan to develop membranes with antioxidant and antimicrobial properties for tissue regeneration (Ciarlantini et al., 2024).
  • Improving hydrogel properties: Research highlighted the benefits of incorporating ethylene glycol dimethacrylate in NVCL-NIPAm hydrogels, enhancing swelling behaviors and mechanical properties for biomedical applications (Tie et al., 2024).
  • Synthesis of high-performance polymers: Ethylene glycol dimethacrylate was utilized to fabricate surface molecularly imprinted polymer membranes for the selective separation and extraction of pharmaceutical compounds, showcasing its role in high-performance polymer synthesis (Zhao et al., 2024).
  • Dual-functional polymer applications: A study reported on the use of ethylene glycol dimethacrylate in the development of dual-functional molecularly imprinted polymers integrated with carbon dots for detecting and adsorbing contaminants in food, demonstrating its versatility in environmental and food safety applications (Wang et al., 2024).
  • Advanced sensing materials: The crosslinking properties of ethylene glycol dimethacrylate were explored in the development of a quartz crystal microbalance sensor, improving its application in food analysis and showcasing its effectiveness in sensor technology (Demir et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 97.5 % (a/a)
Density (d 20 °C/ 4 °C): 1.048 - 1.051
Identity (IR): passes test
The material is stabilised with hydroquinone monomethyl ether.
At time of analytical testing a content of >10 ppm stabiliser was determined.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - Pensky-Martens closed cup

Flash Point(C)

104 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service