Skip to Content
Merck
All Photos(1)

Key Documents

8.18795

Sigma-Aldrich

2-Decanone

for synthesis

Synonym(s):

2-Decanone, Methyl octyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€98.40

€98.40


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
25 ML
€98.40

About This Item

Linear Formula:
CH3(CH2)7COCH3
CAS Number:
Molecular Weight:
156.27
MDL number:
UNSPSC Code:
12352115
EC Index Number:
211-752-6
NACRES:
NA.22

€98.40


Estimated to ship on05 May 2025Details


Request a Bulk Order

Quality Level

Assay

≥95.0% (GC)

form

liquid

mp

3 °C

transition temp

flash point 86 °C

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(CCCCCCCC)C

InChI

1S/C10H20O/c1-3-4-5-6-7-8-9-10(2)11/h3-9H2,1-2H3

InChI key

ZAJNGDIORYACQU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.822 - 0.824
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service