Skip to Content
Merck
All Photos(1)

Key Documents

8.18752

Sigma-Aldrich

3-Chloro-4-fluoroaniline

for synthesis

Synonym(s):

3-Chloro-4-fluoroaniline, chlorofluoroaniline, CFA

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€70.70

€70.70


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€70.70

About This Item

Linear Formula:
3-(Cl),4-(F)C6H3NH2
CAS Number:
Molecular Weight:
145.56
MDL number:
UNSPSC Code:
12352116
EC Index Number:
206-682-8
NACRES:
NA.22

€70.70


Estimated to ship on02 May 2025Details


Request a Bulk Order

vapor pressure

0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

potency

506 mg/kg LD50, oral (Rat)

pH

7 (20 °C, 10 g/L in H2O)

bp

230 °C/1013 hPa

mp

42-45 °C

transition temp

flash point 114 °C

density

1.59 g/cm3

bulk density

1000 kg/m3

storage temp.

2-30°C

SMILES string

Fc1c(cc(cc1)N)Cl

InChI

1S/C6H5ClFN/c7-5-3-4(9)1-2-6(5)8/h1-3H,9H2

InChI key

YSEMCVGMNUUNRK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 42 °C
Melting range (upper value): ≤ 45 °C
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service