Skip to Content
Merck
All Photos(1)

Key Documents

8.18725

Sigma-Aldrich

(2R,3R)-(-)-Di-O-4-toluoyl-L-tartaric acid

for synthesis

Synonym(s):

(2R,3R)-(-)-Di-O-4-toluoyl-L-tartaric acid, Di-O-4-toluoyl-L-tartaric acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€105.00
100 G
€231.00

€105.00


Estimated to ship on17 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€105.00
100 G
€231.00

About This Item

Linear Formula:
(4-(CH3)C6H4COOCHCOOH)2
CAS Number:
Molecular Weight:
386.35
MDL number:
UNSPSC Code:
12352106
EC Index Number:
251-131-7
NACRES:
NA.22

€105.00


Estimated to ship on17 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥99.0% (acidimetric)

form

powder

mp

165-171 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)[C@H](OC(=O)c2ccc(cc2)C)[C@H](OC(=O)c1ccc(cc1)C)C(=O)[O-]

InChI

1S/C20H18O8/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24)/p-2/t15-,16+

InChI key

CMIBUZBMZCBCAT-IYBDPMFKSA-L

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Spec. rotation [α²0/D (c=1 in ethanol): -142.0 - -136.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service