Skip to Content
Merck
All Photos(1)

Key Documents

8.18694

Sigma-Aldrich

Allyl acetoacetate

for synthesis

Synonym(s):

Allyl acetoacetate, Acetoacetic acid allyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€119.00

€119.00


Estimated to ship on13 June 2025Details



Select a Size

Change View
100 ML
€119.00

About This Item

Linear Formula:
CH3COCH2COOCH2CH=CH2
CAS Number:
Molecular Weight:
142.15
MDL number:
UNSPSC Code:
12352108
EC Index Number:
214-269-9
NACRES:
NA.22

€119.00


Estimated to ship on13 June 2025Details


vapor pressure

0.2 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

autoignition temp.

300 °C

potency

259 mg/kg LD50, oral (Rat)
266 mg/kg LD50, skin (Rat)

expl. lim.

1.15 % (v/v)

pH

3.7 (20 °C, 48 g/L in H2O)

bp

87-91 °C/13 hPa

mp

<-70 °C

transition temp

flash point 67 °C

solubility

48 g/L

density

1.038 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC=C)C(=O)CC(=O)C

InChI

1S/C7H10O3/c1-3-4-10-7(9)5-6(2)8/h3H,1,4-5H2,2H3

InChI key

AXLMPTNTPOWPLT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.036 - 1.041
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service