Skip to Content
Merck
All Photos(1)

Key Documents

8.18663

Sigma-Aldrich

5-Methyl-3-heptanone

for synthesis

Synonym(s):

5-Methyl-3-heptanone, Ethyl amyl ketone, Ethyl isoamyl ketone, Isoamyl ethyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€77.70

€77.70


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€77.70

About This Item

Linear Formula:
CH3CH2COCH2CH(CH3)CH2CH3
CAS Number:
Molecular Weight:
128.21
MDL number:
UNSPSC Code:
12352115
EC Index Number:
208-793-7
NACRES:
NA.22

€77.70


Estimated to ship on07 May 2025Details


Request a Bulk Order

vapor pressure

2.7 hPa ( 25 °C)

Quality Level

Assay

≥94.0% (GC)

form

liquid

potency

3500 mg/kg LD50, oral (Rat)
>16000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-48 % (v/v)

bp

157-162 °C/1013 hPa

mp

-57 °C

transition temp

flash point 43 °C

solubility

3 g/L

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(CC(CC)C)CC

InChI

1S/C8H16O/c1-4-7(3)6-8(9)5-2/h7H,4-6H2,1-3H3

InChI key

PSBKJPTZCVYXSD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 94.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.820 - 0.823
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service