Skip to Content
Merck
All Photos(2)

Key Documents

8.18532

Sigma-Aldrich

(1-Methoxy-2-propyl) acetate

(stabilised with 2,6-di-tert-butyl-4-methyl-phenol) for synthesis

Synonym(s):

(1-Methoxy-2-propyl) acetate, Acetic acid 2-methoxypropyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€52.90
2.5 L
€105.00

€52.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€52.90
2.5 L
€105.00

About This Item

Linear Formula:
CH3COOCH(CH3)CH2OCH3
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-603-9
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
145-147 °C/1013 hPa
vapor pressure:
3.1 hPa ( 20 °C)

€52.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

3.1 hPa ( 20 °C)

Assay

≥96.0% (GC)

form

liquid

autoignition temp.

315 °C

expl. lim.

1.5 % (v/v)

dilution

(for synthesis)

pH

4 (20 °C, 200 g/L in H2O)

bp

145-147 °C/1013 hPa

mp

-67 °C

transition temp

flash point 45 °C

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC(OC(=O)C)C)C

InChI

1S/C6H12O3/c1-5(4-8-3)9-6(2)7/h5H,4H2,1-3H3

InChI key

LLHKCFNBLRBOGN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.966 - 0.969
(2-Methoxy-1-propyl) acetate (GC, area%): ≤ 3.0 % (a/a)
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

113.0 °F

Flash Point(C)

45 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service