Skip to Content
Merck
All Photos(1)

Key Documents

8.18469

Sigma-Aldrich

2,4,6-Trichlorophenol

for synthesis

Synonym(s):

2,4,6-Trichlorophenol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€38.10
1 KG
€184.00

€38.10


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€38.10
1 KG
€184.00

About This Item

Empirical Formula (Hill Notation):
C6H3Cl3O
CAS Number:
Molecular Weight:
197.45
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-795-9
NACRES:
NA.22

€38.10


Estimated to ship on14 April 2025Details


Request a Bulk Order

vapor pressure

0.035 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

powder

potency

887 mg/kg LD50, oral (Rat)

bp

244-246 °C/1013 hPa

mp

65-68 °C

transition temp

flash point 99 °C

solubility

0.8 g/L

density

1.675 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

Clc1c(c(cc(c1)Cl)Cl)O

InChI

1S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

LINPIYWFGCPVIE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 65 °C
Melting range (upper value): ≤ 68 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service