Skip to Content
Merck
All Photos(1)

Key Documents

8.18455

Sigma-Aldrich

Eugenol

for synthesis

Synonym(s):

Eugenol, 4-Allyl-2-methoxyphenol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€44.80
500 ML
€152.00

€44.80


Estimated to ship on10 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€44.80
500 ML
€152.00

About This Item

Linear Formula:
2-(CH3O)-4-(CH2=CHCH2)C6H3OH
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-589-1
NACRES:
NA.22

€44.80


Estimated to ship on10 June 2025Details


Request a Bulk Order

vapor pressure

<0.1 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

1930 mg/kg LD50, oral (Rat)

bp

252 °C/1013 hPa

mp

-9.2 °C

transition temp

flash point >110 °C

density

1.07 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)c1c(ccc(c1)CC=C)O

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3,5-7,11H,1,4H2,2H3

InChI key

RRAFCDWBNXTKKO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.064 - 1.066
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service