Skip to Content
Merck
All Photos(2)

Key Documents

8.18410

Sigma-Aldrich

(S)-(+)-Carvone

for synthesis

Synonym(s):

(S)-(+)-Carvone, (S)-(+)-5-Isopropenyl-2-methyl-2-cyclohexenone, (S)-(+)-p-Mentha-6,8-dien-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€51.20

€51.20


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 ML
€51.20

About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
MDL number:
UNSPSC Code:
12352115
EC Index Number:
218-827-2
NACRES:
NA.22

€51.20


In StockDetails


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

3562 mg/kg LD50, oral (Rat)

bp

228-230 °C/1013 hPa

transition temp

flash point 88 °C (DIN 51758)

solubility

29-79 g/L

density

0.96 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C1C[C@H](CC=C1C)C(=C)C

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3/t9-/m0/s1

InChI key

ULDHMXUKGWMISQ-VIFPVBQESA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.958 - 0.962
Spec. rotation [α²0/D (undiluted): +57.0 - +62.0 °
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.1 °F - closed cup

Flash Point(C)

95.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service