Skip to Content
Merck
All Photos(1)

Key Documents

8.18402

Sigma-Aldrich

Polytetrahydrofuran 1000

(stabilised with 2,6-di-tert-butyl-4-methylphenol) for synthesis

Synonym(s):

Polytetrahydrofuran 1000, PTHF

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€33.70
500 G
€73.30

€33.70


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
€33.70
500 G
€73.30

About This Item

Linear Formula:
H(C4H8O)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.21

€33.70


In StockDetails


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

liquid

potency

>5000 mg/kg LD50, oral (Rat)

pH

7 (20 °C in H2O, Aqueous solution)

mp

23-28 °C

transition temp

flash point 240 °C

solubility

<10 g/L

density

0.982 g/cm3 at 30 °C

storage temp.

2-30°C

SMILES string

O([C@H](CO)CC)CCCC

InChI

1S/C8H18O2/c1-3-5-6-10-8(4-2)7-9/h8-9H,3-7H2,1-2H3/t8-/m0/s1

InChI key

BJZYYSAMLOBSDY-QMMMGPOBSA-N

Analysis Note

Melting range (lower value): ≥ 23 °C
Melting range (upper value): ≤ 28 °C
Hydroxyl value: 109 - 115
Average molecular mass: 975 - 1025
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service