Skip to Content
Merck
All Photos(1)

Key Documents

8.18165

Sigma-Aldrich

ß-Nitrostyrene

for synthesis

Synonym(s):

ß-Nitrostyrene, ω-Nitrostyrene, (1-Nitro-2-phenyl)ethylene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€87.80

€87.80


Estimated to ship on02 May 2025Details



Select a Size

Change View
25 G
€87.80

About This Item

Empirical Formula (Hill Notation):
C8H7NO2
CAS Number:
Molecular Weight:
149.15
MDL number:
UNSPSC Code:
12352102
EC Index Number:
203-066-0
NACRES:
NA.22

€87.80


Estimated to ship on02 May 2025Details


Quality Level

Assay

≥98.0% (GC)

autoignition temp.

250 °C

bp

250-260 °C

mp

54-57 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])\C=C\c1ccccc1

InChI

1S/C8H7NO2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H/b7-6+

InChI key

PIAOLBVUVDXHHL-VOTSOKGWSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 54 °C
Melting range (upper value): ≤ 57 °C
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service