Skip to Content
Merck
All Photos(1)

Key Documents

8.17074

SAFC

Butylhydroxytoluene

E 321

Pharma Manufacturing

Synonym(s):

Butylhydroxytoluene

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€108.00
5 KG
€398.00
25 KG
€1,780.00

€108.00


Estimated to ship on19 May 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€108.00
5 KG
€398.00
25 KG
€1,780.00

About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
MDL number:
UNSPSC Code:
12352104
E Number:
E321
EC Index Number:
204-881-4

€108.00


Estimated to ship on19 May 2025Details


Request a Bulk OrderRequest a Sample

vapor pressure

0.02 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

345 °C

bp

265 °C/1013 hPa

mp

69-70 °C

transition temp

flash point 127 °C

density

1.05 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

15-25°C

SMILES string

Oc1c(cc(cc1C(C)(C)C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Application

Butylhydroxytoluene is an antioxidant and is typically used as an additive for the preservation of food.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service