Skip to Content
Merck

8.17058

SAFC

Maleic acid

EMPROVE® ESSENTIAL, Ph. Eur., NF

Pharma Manufacturing

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€137.00
25 KG
€1,830.00

€137.00


Estimated to ship on16 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€137.00
25 KG
€1,830.00

About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein:
605762
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-742-5

€137.00


Estimated to ship on16 April 2025Details


Request a Bulk OrderRequest a Sample

Agency

NF
Ph. Eur.

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

99.0-101.0% substance (anhydrous) basis

form

solid

potency

708 mg/kg LD50, oral (Rat)
1560 mg/kg LD50, skin (Rabbit)

technique(s)

API processing | cocrystal formation: suitable
API processing | salt formation: suitable

pH

1.3 (20 °C, 100 g/L in H2O)

mp

130-135 °C (lit.)

solubility

478.8 g/L

density

1.59 g/mL at 25 °C (lit.)

cation traces

Fe: ≤5 ppm
V: ≤1 ppm
heavy metals (as Pb): ≤10 ppm

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Maleic acid is typically used as pH adjuster or buffer agent in pharmaceutical formulations. Maleic acid Emprove® Essential is mainly used in highly regulated pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service