Skip to Content
Merck
All Photos(1)

Documents

8.17054

SAFC

Sodium gluconate

E 576

Pharma Manufacturing

Synonym(s):

Sodium gluconate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NaO7
CAS Number:
Molecular Weight:
218.14
MDL number:
UNSPSC Code:
12352107
E Number:
E576
EC Index Number:
208-407-7

Quality Level

form

powder

autoignition temp.

>200 °C

potency

>2000 mg/kg LD50, oral (Rat)

pH

7.0-8.0 (20 °C, 100 g/L in H2O)

mp

170-175 °C

solubility

590 g/L

application(s)

pharma/biopharma processes

storage temp.

15-25°C

InChI

1S/C6H12O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1

InChI key

UPMFZISCCZSDND-UHFFFAOYSA-M

Application

Sodium gluconate is typically used as buffering and chelating agent. Sodium gluconate E 576 is suitable for use in pharmaceutical processes and as food additive.

Disclaimer

The substance 8.17054 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service