Skip to Content
Merck

8.17001

SAFC

Polyethylene glycol

200, EMPROVE® ESSENTIAL, DAB 8, NF

Pharma Manufacturing

Synonym(s):

Poly(ethylene glycol), Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 200, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

5 L
€198.00
25 L
Inquire
220 KG
Inquire

€198.00


Estimated to ship on24 March 2025Details


Request a Bulk Order

Select a Size

Change View
5 L
€198.00
25 L
Inquire
220 KG
Inquire

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2

€198.00


Estimated to ship on24 March 2025Details


Request a Bulk Order

Agency

DAB 8
NF

Quality Level

vapor pressure

<0.01 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

liquid

autoignition temp.

350 °C (DIN 51794)

mol wt

average Mn 200

pH

5-7 (20 °C, 100 g/L in H2O)

bp

>150 °C

mp

-50 °C

transition temp

flash point 180 °C

solubility

soluble 70 g/L

density

1.124 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solubility enhancement

storage temp.

no temp limit

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

When working on a pharmaceutical formulation, there are specific considerations to make and challenging hurdles to take before you can successfully launch your final drug product - formulation stability, release kinetics and bioavailability limitations just to name a few. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Polyethylene glycol (PEG) is a multipurpose polymer which can be applied in solid, semi-solid and liquid formulations. Its broad functional spectrum includes e.g. improvement of API solubility, acting as a lubricant for tablet coating, function as a carrier, and many more. Our comprehensive portfolio of PEGs comprises diverse polymer sizes of different molecular weights to meet your specific application and needs.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Medicine for children poses unique formulation challenges compared to adults. Consider developmental physiology and age specifics when designing pharmaceuticals. Quality issues can severely impact patient safety. Therefore, excipient quality, supplier selection, and supply chain security are crucial, particularly for pediatric formulations.

Related Content

Developing formulations specifically for infants and children is increasingly important. To help you master these challenges effectively, we offer an extensive portfolio of well-established high-quality excipients and APIs for pediatric pharmaceutical formulations that are proven in practice.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service