Skip to Content
Merck
All Photos(1)

Key Documents

8.14704

Sigma-Aldrich

Methyl 2,4-dihydroxy-3,6-dimethylbenzoate

for synthesis

Synonym(s):

Methyl 2,4-dihydroxy-3,6-dimethylbenzoate, 3,6-Dimethyl-ß-resorcylic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€131.00

€131.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 G
€131.00

About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352108
EC Index Number:
225-193-0
NACRES:
NA.22

€131.00


In StockDetails


Request a Bulk Order

Quality Level

Assay

≥99.0% (HPLC)

form

crystals

mp

144-146 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)c1c(c(c(cc1C)O)C)O

InChI

1S/C10H12O4/c1-5-4-7(11)6(2)9(12)8(5)10(13)14-3/h4,11-12H,1-3H3

InChI key

UUQHKWMIDYRWHH-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 143 °C
Melting range (upper value): ≤ 146 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service