Skip to Content
Merck
All Photos(1)

Key Documents

8.14573

Sigma-Aldrich

Palladium(II) nitrate dihydrate

40% Pd, for synthesis

Synonym(s):

Palladium(II) nitrate dihydrate (40% Pd)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€81.00
1 G
€129.00

€81.00


Estimated to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 MG
€81.00
1 G
€129.00

About This Item

Linear Formula:
Pd(NO3)2 · 2 H2O
CAS Number:
Molecular Weight:
266.46
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-265-8
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
powder

€81.00


Estimated to ship on01 April 2025Details


Request a Bulk Order

Product Name

Palladium(II) nitrate dihydrate (40% Pd), for synthesis

grade

for synthesis
synthesis grade

Quality Level

form

powder

potency

200-2000 mg/kg LD50, oral (Rat)

pH

1 (20 °C, 1 g/L in H2O)

storage temp.

2-30°C

SMILES string

[Pd](O[N+](=O)[O-])O[N+](=O)[O-].O.O

InChI

1S/2NO3.2H2O.Pd/c2*2-1(3)4;;;/h;;2*1H2;/q2*-1;;;+2

InChI key

JUBNUQXDQDMSKL-UHFFFAOYSA-N

Analysis Note

Assay (ICP-OES): ≥ 99.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service