Skip to Content
Merck
All Photos(2)

Key Documents

8.14422

Sigma-Aldrich

N-(Hydroxymethyl)-phthalimide

for synthesis

Synonym(s):

N-(Hydroxymethyl)-phthalimide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€59.30

€59.30


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€59.30

About This Item

Empirical Formula (Hill Notation):
C9H7NO3
CAS Number:
Molecular Weight:
177.16
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-241-4
NACRES:
NA.22

€59.30


Estimated to ship on17 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥99.0% (HPLC)

form

powder

mp

140-146 °C

storage temp.

2-30°C

SMILES string

[n]1([c](c2c([c]1=O)cccc2)=O)CO

InChI

1S/C9H7NO3/c11-5-10-8(12)6-3-1-2-4-7(6)9(10)13/h1-4,11H,5H2

InChI key

MNSGOOCAMMSKGI-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 140 °C
Melting range (upper value): ≤ 146 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service