Skip to Content
Merck
All Photos(1)

Key Documents

8.14254

Sigma-Aldrich

Levulinic acid

for synthesis

Synonym(s):

Levulinic acid, 4-Oxopentanoic acid, 4-Oxovalerianic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€33.10
500 G
€97.70

€33.10


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€33.10
500 G
€97.70

About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-649-2
NACRES:
NA.22

€33.10


Estimated to ship on16 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

solid

potency

1850 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

245-246 °C/1013 mbar

mp

26-29 °C

transition temp

flash point 98 °C

solubility

675 g/L

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCC(=O)C

InChI

1S/C5H8O3/c1-4(6)2-3-5(7)8/h2-3H2,1H3,(H,7,8)

InChI key

JOOXCMJARBKPKM-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service