Skip to Content
Merck
All Photos(1)

Key Documents

8.14109

Sigma-Aldrich

Tributyltin hydride (stabilized)

for synthesis

Synonym(s):

Tributyltin hydride (stabilized), Tributylstannane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€36.70
25 ML
€97.10
100 ML
€291.00

€36.70


Estimated to ship on02 June 2025Details


Request a Bulk Order

Select a Size

Change View
5 ML
€36.70
25 ML
€97.10
100 ML
€291.00

About This Item

Empirical Formula (Hill Notation):
C12H28Sn
CAS Number:
Molecular Weight:
291.06
MDL number:
UNSPSC Code:
12352103
EC Index Number:
211-704-4
NACRES:
NA.22

€36.70


Estimated to ship on02 June 2025Details


Request a Bulk Order

vapor pressure

5 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

>250 °C

reaction suitability

reagent type: reductant

bp

112-114 °C/11 hPa

transition temp

flash point 109 °C

density

1.10 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/3C4H9.Sn/c3*1-3-4-2;/h3*1,3-4H2,2H3;

InChI key

PIILXFBHQILWPS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.100 - 1.102
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service