Skip to Content
Merck
All Photos(1)

Documents

81070N

Sigma-Aldrich

Probumin® Bovine Serum Albumin Manufacturing Grade, 30% Solution

29-31 g/dL protein (Biuret), USA origin, Tested for IgG, enzyme immunoassay: suitable, Serology: suitable, Blood banking: suitable

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41141801
NACRES:
NA.47

product name

Probumin® Bovine Serum Albumin Manufacturing Grade, 30% Solution, pack of 50 mL

description

0.1% sodium azide as preservtive
Contains sodium azide less than 0.1 g/dL

Quality Level

Assay

29-31 g/dL protein basis (biuret)

form

solution

shelf life

36 mo.

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

pack of 50 mL

manufacturer/tradename

Probumin®

origin

USA origin

concentration

29-31 g/dL protein (Biuret)

technique(s)

blood typing: suitable
enzyme immunoassay: suitable

dilution

(Immunohematology)
(Serology)

impurities

Tested for IgG

pH

7.2-7.4

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service