Skip to Content
Merck
All Photos(1)

Key Documents

810653

Sigma-Aldrich

Probumin® Bovine Serum Albumin Microbiological Grade

≥98%, microbiological culture: suitable, 0-5.0% moisture (Karl Fischer), USA origin

Synonym(s):

Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V

Sign Into View Organizational & Contract Pricing

Select a Size

5 KG
€3,380.00

€3,380.00


Estimated to ship on19 May 2025Details


Request a Bulk Order

Select a Size

Change View
5 KG
€3,380.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801

€3,380.00


Estimated to ship on19 May 2025Details


Request a Bulk Order

Product Name

Probumin® Bovine Serum Albumin Microbiological Grade, pack of 5 kg

biological source

bovine serum

Quality Level

Assay

≥98% (purity)

form

lyophilized powder

shelf life

48 mo.

mol wt

apparent mol wt 66 kDa

purified by

heat shock fractionation

packaging

pack of 5 kg

manufacturer/tradename

Probumin®

origin

USA origin

technique(s)

microbiological culture: suitable

impurities

0-5% moisture (Karl Fischer)

loss

≤5%

pH range

6.8-7.2

anion traces

chloride (Cl-): 0.0-12.0 mg/g

cation traces

Na: 0.0-10.0 mg/g

foreign activity

Bluetongue, none detected
Vesicular Stomatitis Virus, none detected

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Manufactured by a proprietary heat-shock fractionation process, using caprylic acid as an albumin stabilizer. This BSA powder is solvent treated and extensively diafiltered. This bovine albumin product is low in sodium and purity is >=98%; typical purity is 100%.

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Probumin® BSA grades are distinguished by differences in production. The use of terms such as ′Diagnostic Grade′ or ′Vaccine Grade′ in product names solely indicates that said products are commonly used by customers in those applications. We have not qualified every single product in every conceivable application. Users should qualify Probumin® BSA grades using the specific reagents, conditions, and constraints of their assay.
For further manufacturing use only. Not for human or animal consumption.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service