Skip to Content
Merck
All Photos(1)

Key Documents

8.08634

Sigma-Aldrich

Ethyl tridecanoate

for synthesis

Synonym(s):

Ethyl tridecanoate, Tridecanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€112.00

€112.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 ML
€112.00

About This Item

Linear Formula:
CH3(CH2)11COOC2H5
CAS Number:
Molecular Weight:
242.40
MDL number:
UNSPSC Code:
12352108
EC Index Number:
248-929-2
NACRES:
NA.22

€112.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)CCCCCCCCCCCC

InChI

1S/C15H30O2/c1-3-5-6-7-8-9-10-11-12-13-14-15(16)17-4-2/h3-14H2,1-2H3

InChI key

QJYYMNOTJXIOBP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.860 - 0.864
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service