Skip to Content
Merck
All Photos(1)

Key Documents

8.08222

Sigma-Aldrich

Trichloroacetonitrile

for synthesis

Synonym(s):

Trichloroacetonitrile, Trichloromethylnitrile

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€93.30
250 ML
€185.00

€93.30


Estimated to ship on11 June 2025Details


Request a Bulk Order

Select a Size

Change View
50 ML
€93.30
250 ML
€185.00

About This Item

Empirical Formula (Hill Notation):
C2Cl3N
CAS Number:
Molecular Weight:
144.39
MDL number:
UNSPSC Code:
12352117
EC Index Number:
208-885-7
NACRES:
NA.22

€93.30


Estimated to ship on11 June 2025Details


Request a Bulk Order

vapor pressure

77 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

250 mg/kg LD50, oral (Rat)

bp

85.7 °C/1013 hPa

mp

-44 °C

transition temp

flash point >195 °C

solubility

750 mg/L

density

1.44 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(Cl)(Cl)C#N

InChI

1S/C2Cl3N/c3-2(4,5)1-6

InChI key

DRUIESSIVFYOMK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.442 - 1.448
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service