Skip to Content
Merck
All Photos(2)

Key Documents

8.08125

Sigma-Aldrich

4,4′-Bis(dimethylamino)-benzophenone

for synthesis

Synonym(s):

4,4′-Bis(dimethylamino)-benzophenone, N,N,N,N′-Tetramethyl-4,4′-diaminobenzophenone, Michler ketone

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€76.50
1 KG
€241.00

€76.50


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€76.50
1 KG
€241.00

About This Item

Linear Formula:
[(CH3)2N]C6H4COC6H4-4-[N(CH3)2]
CAS Number:
Molecular Weight:
268.35
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-027-5
NACRES:
NA.22

€76.50


Estimated to ship on02 May 2025Details


Request a Bulk Order

Quality Level

form

powder

autoignition temp.

480 °C

potency

>6400 mg/kg LD50, oral (Rat)

mp

172-175 °C

transition temp

flash point 220 °C

solubility

0.4 g/L

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

N(C)(C)c1ccc(cc1)C(=O)c2ccc(cc2)N(C)C

InChI

1S/C17H20N2O/c1-18(2)15-9-5-13(6-10-15)17(20)14-7-11-16(12-8-14)19(3)4/h5-12H,1-4H3

InChI key

VVBLNCFGVYUYGU-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 97.0 %
Melting range (lower value): ≥ 171 °C
Melting range (upper value): ≤ 175 °C
Identity (IR): passes test

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service