Skip to Content
Merck
All Photos(1)

Documents

8.07837

Sigma-Aldrich

Antimony(V) chloride

for synthesis

Synonym(s):

Antimony(V) chloride, Antimony pentachloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl5Sb
CAS Number:
Molecular Weight:
299.03
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-601-8
NACRES:
NA.22

vapor pressure

1.1 hPa ( 20 °C)

Quality Level

form

liquid

potency

1115 mg/kg LD50, oral (Rat)

mp

3-4 °C

density

2.35 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/5Cl.Sb

InChI key

WCEPFCFFCMELSL-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service