Skip to Content
Merck
All Photos(1)

Key Documents

8.07480

Sigma-Aldrich

Ethylene glycol monoisopropyl ether

for synthesis

Synonym(s):

Ethylene glycol monoisopropyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€34.40
20 KG
€316.00

€34.40


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 L
€34.40
20 KG
€316.00

About This Item

Empirical Formula (Hill Notation):
C5H12O2
CAS Number:
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-685-6
NACRES:
NA.22

€34.40


In StockDetails


Request a Bulk Order

vapor pressure

5.99 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

240 °C

potency

5111 mg/kg LD50, oral (Rat)
1445 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6-13.0 % (v/v)

mp

<-60 °C

transition temp

flash point 43-45 °C

density

0.90 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(C)C)CCO

InChI

1S/C5H12O2/c1-5(2)7-4-3-6/h5-6H,3-4H2,1-2H3

InChI key

HCGFUIQPSOCUHI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.903 - 0.904
Water (K. F.): ≤ 0.30 %
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

109.4 - 113.0 °F - Equilibrium method

Flash Point(C)

43 - 45 °C - Equilibrium method


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service