Skip to Content
Merck
All Photos(1)

Key Documents

8.07325

Sigma-Aldrich

Piperazine anhydrous

for synthesis

Synonym(s):

Piperazine anhydrous, Diethylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€28.60
250 G
€36.10
1 KG
€123.00
5 KG
€273.00
25 KG
€1,800.00

€28.60


Estimated to ship on02 June 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
€28.60
250 G
€36.10
1 KG
€123.00
5 KG
€273.00
25 KG
€1,800.00

About This Item

Empirical Formula (Hill Notation):
C4H10N2
CAS Number:
Molecular Weight:
86.14
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-808-3
NACRES:
NA.22

€28.60


Estimated to ship on02 June 2025Details


Request a Bulk Order

grade

anhydrous

Quality Level

vapor pressure

15 hPa ( 50 °C)

Assay

≥99.0% (GC)

form

crystals

potency

2600 mg/kg LD50, oral (Rat)
8300 mg/kg LD50, skin (Rabbit)

pH

12 (20 °C, 150 g/L in H2O)

mp

107-111 °C

density

1.1 g/cm3 at 20 °C

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

N1CCNCC1

InChI

1S/C4H10N2/c1-2-6-4-3-5-1/h5-6H,1-4H2

InChI key

GLUUGHFHXGJENI-UHFFFAOYSA-N

Application



  • Enhancement of Polyamide Nanofiltration Membranes: Piperazine anhydrous was utilized to refine polyamide nanofiltration membranes, focusing on reducing pore size distribution through a one-step multiple interfacial polymerization technique. This method significantly enhances the membrane′s antifouling capabilities, important for applications in water treatment and chemical separation (Liu et al., 2022).


  • Ultrasonic synthesis of stable oil filled microcapsules: This study explores the ultrasonic synthesis of oil-filled microcapsules using thiolated chitosan, potentially involving derivatives like ᴅʟ-N-Acetylhomocysteine thiolactone for improved stability and functional properties. The research provides insights into advanced microencapsulation techniques relevant in pharmaceutical and industrial applications (Source).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 109 °C
Melting range (upper value): ≤ 112 °C
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Repr. 2 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service