Skip to Content
Merck
All Photos(1)

Key Documents

8.07107

Sigma-Aldrich

Palladium(II) oxide

85% Pd, hydrogenation catalyst for synthesis

Synonym(s):

Palladium(II) oxide (85% Pd)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€152.00

€152.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 MG
€152.00

About This Item

Empirical Formula (Hill Notation):
OPd
CAS Number:
Molecular Weight:
122.42
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-218-3
NACRES:
NA.22

€152.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Product Name

Palladium(II) oxide (85% Pd), hydrogenation catalyst for synthesis

Quality Level

form

powder

reaction suitability

reagent type: catalyst
reaction type: Hydrogenations

storage temp.

2-30°C

SMILES string

[Pd+2].[O-2]

InChI

1S/O.Pd/q-2;+2

InChI key

JQPTYAILLJKUCY-UHFFFAOYSA-N

General description

hydrogenation catalyst for synthesis

Analysis Note

Assay Pd: ≥ 85.0 %

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service