Skip to Content
Merck
All Photos(1)

Key Documents

8.06770

Sigma-Aldrich

Nitrobenzene

for synthesis

Synonym(s):

Nitrobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€31.40
1 L
€50.90
2.5 L
€87.30
33 KG
€537.00

€31.40


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€31.40
1 L
€50.90
2.5 L
€87.30
33 KG
€537.00

About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
MDL number:
UNSPSC Code:
12352102
EC Index Number:
202-716-0
NACRES:
NA.22

€31.40


Estimated to ship on07 May 2025Details


Request a Bulk Order

vapor pressure

0.3 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

480 °C (DIN 51794)

potency

640 mg/kg LD50, oral (Rat)
2100 mg/kg LD50, skin (Rat)

expl. lim.

1.8-40 % (v/v)

refractive index

n20/589 1.553

pH

8.1 (20 °C, 1 g/L in H2O)

bp

211 °C/1013 hPa

mp

6.0 °C

transition temp

flash point 88 °C

solubility

1.90 g/L

density

1.20 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1ccccc1

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

LQNUZADURLCDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 1.201 - 1.203
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 1B - STOT RE 1 Inhalation

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service